Merck
  • Diosmin induces genotoxicity and apoptosis in DU145 prostate cancer cell line.
Diosmin induces genotoxicity and apoptosis in DU145 prostate cancer cell line.
Toxicology in vitro : an international journal published in association with BIBRA (2014-12-17)
Anna Lewinska, Justyna Siwak, Iwona Rzeszutek, Maciej Wnuk
ABSTRACT
Plant-derived dietary polyphenolic compounds, such as flavonoids, with cancer cell-specific pro-apoptotic activity and chemopreventive potential are thought to be promising anticancer agents. In the present study, we were interested in determining if flavonoid-induced genotoxicity may also provoke cancer cell death. Cyto- and genotoxicity of three selected flavonoid glycosides (naringin, diosmin and hesperidin) in DU145 prostate cancer cell line were investigated. Flavonoid glycosides decreased cancer cell number and proliferative activity in a different manner. Flavonoid glycosides induced oxidative stress: intracellular total ROS and superoxide production were augmented after flavonoid treatment. Flavonoid glycosides stimulated DNA double strand breaks (DSBs) and micronuclei production. Diosmin was found the most potent genotoxic agent in DU145 cells, which, in turn, resulted in its pro-apoptotic activity. The more robust recruitment of 53BP1 was correlated with lower DNA and chromosomal damage after naringin and hesperidin treatment compared with diosmin treatment. Flavonoid glycosides were also found to be DNA hypomethylating agents with an ability to modulate cancer cell epigenome leading to changes in the gene expression patterns. Taken together, diosmin, a dietary flavonoid glycoside, was found active against DU145 cells by promoting genotoxic events and a concomitant apoptotic cell death. Thus, a comprehensive analysis of biological activity of diosmin against cancer cells both in vitro and in vivo deserves further investigation.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
2′,7′-Dichlorofluorescein, ACS reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Naringin, ≥95% (HPLC)
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Hesperidin, ≥80% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Naringin, ≥90% (HPLC), from citrus fruit
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
2′,7′-Dichlorofluorescein, suitable for use as an indicator in chloride titration, ~90% (TLC), powder
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Supelco
Hesperidin, analytical standard
Supelco
Naringin, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
2′,7′-Dichlorofluorescein, BioReagent, suitable for fluorescence, ≥90% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%