Merck
  • Tyrosine nitration in peptides by peroxynitrite generated in situ in a light-controlled platform: effects of pH and thiols.
Tyrosine nitration in peptides by peroxynitrite generated in situ in a light-controlled platform: effects of pH and thiols.
Journal of inorganic biochemistry (2014-05-27)
Tara R deBoer, Rafael I Palomino, Sharon O Idiga, Glenn L Millhauser, Pradip K Mascharak
ABSTRACT
Peroxynitrite has been shown to play a critical role in inflammation and affords 3-nitrotyrosine as the hallmark product. The reported methods of generating this reactive nitrogen species in situ often fails to provide a high and steady flux of peroxynitrite resulting in poor yields of 3-nitrotyrosine. Herein we report a two-component peroxynitrite-generating platform in which this anion is produced in a biomimetic fashion and under the control of visible light. Incorporation of the nitric oxide- and superoxide-generating components in polymer matrices allows easy alterations of pH in the reaction wells of this platform. We have demonstrated very efficient nitration of tyrosine by peroxynitrite at different pH values and with varying concentrations of carbonate. In addition to tyrosine, a set of tyrosine-containing peptides was also studied. Presence of glutathione in the reaction wells increases the extent of tyrosine nitration in such peptide substrates presumably by raising the lifetime of nitric oxide in the reaction medium. When a cysteine residue was included in the sequence of the peptide, the extent of nitration of the tyrosine residue was found to depend on the position of the cysteine residue with respect to tyrosine. The extent of tyrosine nitration is strongly attenuated when the cysteine residue is directly adjacent to the tyrosine. This effect has been attributed to an intramolecular radical transfer mechanism. Taken together, results of this study demonstrate the potential of this light-controlled platform as a convenient bioanalytical tool in studying the reactions of peroxynitrite under widely varying conditions.
Sigma-Aldrich
Xanthine, BioUltra, ≥99%
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
Xanthine, ≥99%
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hypoxanthine, ≥99.0%