Merck
  • Spirodela polyrhiza stimulates the growth of its endophytes but differentially increases their fenpropathrin-degradation capabilities.
Spirodela polyrhiza stimulates the growth of its endophytes but differentially increases their fenpropathrin-degradation capabilities.
Chemosphere (2015-02-07)
Xing-Jian Xu, Ji-Quan Sun, Yong Nie, Xiao-Lei Wu
ABSTRACT
In situ remediation of organic contaminants via physical, chemical, and biological approaches is a practical technique for cleansing contaminated water and soil. In the present study, we showed that the three bacterial strains Pseudomonas sp. E1, Klebsiella terrigena E42, and Pseudomonas sp. E46, which can infect and colonize the aquatic plant Spirodela polyrhiza, utilize fenpropathrin as the sole carbon source for growth. S. polyrhiza helped enhance fenpropathrin degradation by E46 by 17.5%, only slightly improved fenpropathrin degradation by E42, and had no effect on strain E1. The application of plant exudates and extracts from fenpropathrin-unexposed/induced plants stimulated bacterial growth of the three strains, but resulted in differential fenpropathrin degradation, suggesting that not all plants and their endophytic bacteria are suitable for coupling phytoremediation and microbial-remediation. Moreover, addition of soil sediments to a microcosm not only stimulated the growth of strain E46 but also increased the rate of fenpropathrin degradation.
Product Description
Supelco
Fenpropathrin, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard