Merck
  • Malarial anemia: digestive vacuole of Plasmodium falciparum mediates complement deposition on bystander cells to provoke hemophagocytosis.
Malarial anemia: digestive vacuole of Plasmodium falciparum mediates complement deposition on bystander cells to provoke hemophagocytosis.
Medical microbiology and immunology (2014-07-06)
Prasad Dasari, Anja Fries, Sophia D Heber, Abdulgabar Salama, Igor-Wolfgang Blau, Klaus Lingelbach, Sebastian Chakrit Bhakdi, Rachanee Udomsangpetch, Michael Torzewski, Karina Reiss, Sucharit Bhakdi
ABSTRACT
The digestive vacuole (DV) of Plasmodium falciparum, which is released into the bloodstream upon rupture of each parasitized red blood cell (RBC), was recently discovered to activate the alternative complement pathway. In the present work, we show that C3- and C5-convertases assembling on the parasitic organelle are able to provoke deposition of activated C3 and C5b-9 on non-infected bystander erythrocytes. Direct contact of DVs with cells is mandatory for the effect, and bystander complement deposition occurs focally, possibly at the sites of contact. Complement opsonization promotes protracted erythrophagocytosis by human macrophages, an effect that is magnified when ring-stage infected RBCs with reduced CD55 and CD59, or paroxysmal nocturnal hemoglobinuria (PNH)-RBCs lacking these complement inhibitors are employed as targets. Bystander attack can also directly induce lysis of PNH-RBCs. Direct evidence for complement activation and bystander attack mediated by DVs was obtained through immunohistochemical analyses of brain paraffin sections from autopsies of patients who had died of cerebral malaria. C3d and the assembled C5b-9 complex could be detected in all sections, colocalizing with and often extending locally beyond massive accumulations of DVs that were identified under polarized light. This is the first demonstration that a complement-activating particle can mediate opsonization of bystander cells to promote their antibody-independent phagocytosis. The phenomenon may act in concert with other pathomechanisms to promote the development of anemia in patients with severe malaria.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C