Merck
  • Properties and extraction of pectin-enriched materials from sugar beet pulp by ultrasonic-assisted treatment combined with subcritical water.
Properties and extraction of pectin-enriched materials from sugar beet pulp by ultrasonic-assisted treatment combined with subcritical water.
Food chemistry (2014-08-31)
Hai-ming Chen, Xiong Fu, Zhi-gang Luo
ABSTRACT
Pectin-enriched material (PEM) was extracted from sugar beet pulp using subcritical water combined with ultrasonic-assisted treatment. Optimisation of the reaction parameters for maximum extraction yield of PEM was carried out using response surface methodology. Optimum modification conditions were as follows: liquid/solid ratio 44.03, extraction temperature 120.72°C, extraction time 30.49min and extraction pressure 10.70MPa. Under optimal conditions, the maximum yield of PEM was 24.63%. The composition of the PEM was determined. The data showed that the contents of galacturonic acid and arabinose were 59.12% and 21.66%, respectively. The flow behaviours were investigated by a rheometer. The effects of PEM on the pasting and thermal properties of maize starch were also conducted. The results showed that the addition of PEM increased pasting temperature and decreased other pasting parameters. Increasing PEM concentrations resulted in increased gelatinisation temperature and enthalpy.
Sigma-Aldrich
L-(−)-Fucose, ≥98% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Millipore
myo-Inositol, ≥99.0%, suitable for microbiology
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
L-(+)-Arabinose, 99%
Sigma-Aldrich
L-(+)-Arabinose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
L-(+)-Arabinose, 98%
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
myo-Inositol, ≥99%
Sigma-Aldrich
myo-Inositol, ≥99% (GC), BioReagent
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%