Merck
  • Shiga-like toxin B subunit of Escherichia coli as scaffold for high-avidity display of anti-immunocomplex peptides.
Shiga-like toxin B subunit of Escherichia coli as scaffold for high-avidity display of anti-immunocomplex peptides.
Analytical chemistry (2014-05-07)
Gabriel Lassabe, Martín Rossotti, Andrés González-Techera, Gualberto González-Sapienza
ABSTRACT
Small compounds cannot bind simultaneously to two antibodies, and thus, their immunodetection is limited to competitive formats in which the analyte is indirectly quantitated by measuring the unoccupied antibody binding sites using a competing reporter. This limitation can be circumvented by using phage-borne peptides selected for their ability to specifically react with the analyte-antibody immunocomplex, which allows the detection of these small molecules in a noncompetitive format (PHAIA) with increased sensitivity and a positive readout. In an effort to find substitutes for the phage particles in PHAIA, we explore the use of the B subunit of the Shiga-like toxin of Escherichia coli, also known as verotoxin (VTX), as a scaffold for multivalent display of anti-immunocomplex peptides. Using the herbicides molinate and clomazone as model compounds, we built peptide-VTX recombinant chimeras that were produced in the periplasmic space of E. coli as soluble pentamers, as confirmed by multiangle light scattering analysis. These multivalent constructs, which we termed nanopeptamers, were conjugated to a tracer enzyme and used to detect the herbicide-antibody complex in an ELISA format. The VTX-nanopeptamer assays performed with over a 10-fold increased sensitivity and excellent recovery from spiked surface and mineral water samples. The carbon black-labeled peptide-VTX nanopeptamers showed great potential for the development of a lateral-flow test for small molecules with a visual positive readout that allowed the detection of up to 2.5 ng/mL of clomazone.
Product Description
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
Supelco
Clomazone, PESTANAL®, analytical standard
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Supelco
Molinate, PESTANAL®, analytical standard
SAFC
Isopropyl β-D-1-thiogalactopyranoside