Merck
Insights into the lysine acetylproteome of human sperm.
Journal of proteomics (2014-07-20)
Guohai Sun, Min Jiang, Tao Zhou, Yueshuai Guo, Yiqiang Cui, Xuejiang Guo, Jiahao Sha
ABSTRACT
Protein lysine acetylation is a dynamic and reversible post-modification that is known to play diverse functions in eukaryotes. Nevertheless, the composition and function of non-histone lysine acetylation in gametes remain unknown. In humans, only capacitated sperm have the capacity to fertilize an egg. In the present study, we found complex composition of lysine acetylated proteins in capacitated human sperm. In vitro fertilization inhibition assay by anti-acetyllysine antibody showed essential roles of lysine acetylation in fertilization. And inhibition of lysine deacetylases, the histone deacetylases, by trichostatin A and nicotinamide, could significantly suppress sperm motility. After immunopurification enrichment of acetylpeptides with anti-acetyllysine antibody and high-throughput liquid chromatography-tandem mass spectrometry identification, we characterized 1206 lysine acetylated sites, corresponding to 576 lysine acetylated proteins in human capacitated sperm. Bioinformatics analysis showed that these proteins are associated with sperm functions, including motility, capacitation, acrosome reaction and sperm-egg interaction. Thus, lysine acetylation is expected to be an important regulatory mechanism for sperm functions. And our characterization of lysine acetylproteome could be a rich resource for the study of male fertility. Mature sperm are almost transcriptionally and translationally silent, thus post-translational modifications play important roles in sperm functions. Till now, only two types of PTMs, phosphorylation and glycosylation, are well studied in normal human sperm based on large scale proteomics. In the present study, we established the acetylproteome of capacitated human sperm. Over 1000 lysine acetylated sites were identified. Bioinformatics analysis shows that lysine acetylated proteins participate in many biological events of sperm functions. We further provided functional data that the lysine acetylation is essential for sperm motility and fertilization using histone acetylase inhibitors and anti-acetyllysine antibody. These data can be strong evidences for the important function of lysine acetylation in human sperm.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Sodium DL-lactate solution, synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)
Sigma-Aldrich
Sodium DL-lactate solution, syrup, BioReagent, 60 % (w/w), synthetic, suitable for cell culture
Sigma-Aldrich
Sodium DL-lactate solution, syrup, 60 % (w/w), synthetic
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Sodium DL-lactate, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%