Merck
  • Lipopolysaccharide augments the uptake of oxidized LDL by up-regulating lectin-like oxidized LDL receptor-1 in macrophages.
Lipopolysaccharide augments the uptake of oxidized LDL by up-regulating lectin-like oxidized LDL receptor-1 in macrophages.
Molecular and cellular biochemistry (2014-10-29)
Ekhtear Hossain, Akinobu Ota, Sivasundaram Karnan, Miyuki Takahashi, Shahnewaj B Mannan, Hiroyuki Konishi, Yoshitaka Hosokawa
ABSTRACT
There is a growing body of evidence supporting an intimate association of immune activation with the pathogenesis of cardiovascular diseases, including atherosclerosis. Uptake of oxidized low-density lipoprotein (oxLDL) through scavenging receptors promotes the formation of mature lipid-laden macrophages, which subsequently leads to exacerbation of regional inflammation and atherosclerotic plaque formation. In this study, we first examined changes in the mRNA level of the lectin-like oxLDL receptor-1 (LOX-1) in the mouse macrophage cell line RAW264.7 and the human PMA-induced macrophage cell line THP-1 after LPS stimulation. LPS significantly up-regulated LOX-1 mRNA in RAW264.7 cells; LOX-1 cell-surface protein expression was also increased. Flow cytometry and fluorescence microscopy analyses showed that cellular uptake of fluorescence (Dil)-labeled oxLDL was significantly augmented with LPS stimulation. The augmented uptake of Dil-oxLDL was almost completely abrogated by treatment with an anti-LOX-1 antibody. Of note, knockdown of Erk1/2 resulted in a significant reduction of LPS-induced LOX-1 up-regulation. Treatment with U0126, a specific inhibitor of MEK, significantly suppressed LPS-induced expression of LOX-1 at both the mRNA and protein levels. Furthermore, LOX-1 promoter activity was significantly augmented by LPS stimulation; this augmentation was prevented by U0126 treatment. Similar results were also observed in human PMA-induced THP-1 macrophages. Taken together, our results indicate that LPS up-regulates LOX-1, at least in part through activation of the Erk1/2 signaling pathway, followed by augmented cellular oxLDL uptake, thus highlighting a critical role of TLR4-mediated aberrant LOX-1 signaling in the pathogenesis of atherosclerosis.
Product Description
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Supelco
Methyl Ethyl Ketone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
Caffeic acid phenethyl ester, ≥97% (HPLC), powder
Supelco
2-Butanone, analytical standard
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard