Merck
  • Microstructural analysis of the fast gelling freeze-dried sodium hyaluronate.
Microstructural analysis of the fast gelling freeze-dried sodium hyaluronate.
Journal of pharmaceutical and biomedical analysis (2014-12-05)
Andrea Krüger-Szabó, Zoltán Aigner, Emese Balogh, István Sebe, Romána Zelkó, István Antal
ABSTRACT
Although sodium hyaluronate is a very unstable and heat sensitive molecule, it can remain relatively stable during the freeze-drying process. Aqueous sodium hyaluronate (NaHA) gels were prepared and the obtained samples were freeze-dried. The freeze-dried NaHA samples showed fast gelling ability meanwhile preserved their initial viscoelasticity even after reconstitution. The microstructure of gels obtained from raw substance and freeze-dried NaHA samples was characterized with positron annihilation lifetime spectroscopy and X-ray diffraction patterns while their functionality-related macrostructural properties were tested based on their rheological behavior. The presence of phosphate salts improved the formation of ordered supramolecular structure retaining water in the free volume holes of the polymer chains characterized with decreased ortho-positronium lifetime values. This property may be advantageous in the development of a freeze-dried NaHA injection dosage form.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard