Merck
  • The cardiac ryanodine receptor luminal Ca2+ sensor governs Ca2+ waves, ventricular tachyarrhythmias and cardiac hypertrophy in calsequestrin-null mice.
The cardiac ryanodine receptor luminal Ca2+ sensor governs Ca2+ waves, ventricular tachyarrhythmias and cardiac hypertrophy in calsequestrin-null mice.
The Biochemical journal (2014-04-25)
Jingqun Zhang, Biyi Chen, Xiaowei Zhong, Tao Mi, Ang Guo, Qiang Zhou, Zhen Tan, Guogen Wu, Alexander W Chen, Michael Fill, Long-Sheng Song, S R Wayne Chen
ABSTRACT
CASQ2 (cardiac calsequestrin) is commonly believed to serve as the SR (sarcoplasmic reticulum) luminal Ca2+ sensor. Ablation of CASQ2 promotes SCWs (spontaneous Ca2+ waves) and CPVT (catecholaminergic polymorphic ventricular tachycardia) upon stress but not at rest. How SCWs and CPVT are triggered by stress in the absence of the CASQ2-based luminal Ca2+ sensor is an important unresolved question. In the present study, we assessed the role of the newly identified RyR2 (ryanodine receptor 2)-resident luminal Ca2+ sensor in determining SCW propensity, CPVT susceptibility and cardiac hypertrophy in Casq2-KO (knockout) mice. We crossbred Casq2-KO mice with RyR2 mutant (E4872Q+/-) mice, which lack RyR2-resident SR luminal Ca2+ sensing, to generate animals with both deficiencies. Casq2+/- and Casq2-/- mice showed stress-induced VTs (ventricular tachyarrhythmias), whereas Casq2+/-/E4872Q+/- and Casq2-/-/E4872Q+/- mice displayed little or no stress-induced VTs. Confocal Ca2+ imaging revealed that Casq2-/- hearts frequently exhibited SCWs after extracellular Ca2+ elevation or adrenergic stimulation, whereas Casq2-/-/E4872Q+/- hearts had few or no SCWs under the same conditions. Cardiac hypertrophy developed and CPVT susceptibility increased with age in Casq2-/- mice, but not in Casq2-/-/E4872Q+/- mice. However, the amplitudes and dynamics of voltage-induced Ca2+ transients in Casq2-/- and Casq2-/-/E4872Q+/- hearts were not significantly different. Our results indicate that SCWs, CPVT and hypertrophy in Casq2-null cardiac muscle are governed by the RyR2-resident luminal Ca2+ sensor. This implies that defects in CASQ2-based lumi-nal Ca2+ sensing can be overridden by the RyR2-resident luminal Ca2+ sensor. This makes this RyR2-resident sensor a promising molecular target for the treatment of Ca2+-mediated arrhythmias.
Product Description
Supelco
2,3-Butanedione, analytical standard
Sigma-Aldrich
2,3-Butanedione, 97%