Merck
  • Silane-catalysed fast growth of large single-crystalline graphene on hexagonal boron nitride.
Silane-catalysed fast growth of large single-crystalline graphene on hexagonal boron nitride.
Nature communications (2015-03-12)
Shujie Tang, Haomin Wang, Hui Shan Wang, Qiujuan Sun, Xiuyun Zhang, Chunxiao Cong, Hong Xie, Xiaoyu Liu, Xiaohao Zhou, Fuqiang Huang, Xiaoshuang Chen, Ting Yu, Feng Ding, Xiaoming Xie, Mianheng Jiang
ABSTRACT
The direct growth of high-quality, large single-crystalline domains of graphene on a dielectric substrate is of vital importance for applications in electronics and optoelectronics. Traditionally, graphene domains grown on dielectrics are typically only ~1 μm with a growth rate of ~1 nm min(-1) or less, the main reason is the lack of a catalyst. Here we show that silane, serving as a gaseous catalyst, is able to boost the graphene growth rate to ~1 μm min(-1), thereby promoting graphene domains up to 20 μm in size to be synthesized via chemical vapour deposition (CVD) on hexagonal boron nitride (h-BN). Hall measurements show that the mobility of the sample reaches 20,000 cm(2) V(-1) s(-1) at room temperature, which is among the best for CVD-grown graphene. Combining the advantages of both catalytic CVD and the ultra-flat dielectric substrate, gaseous catalyst-assisted CVD paves the way for synthesizing high-quality graphene for device applications while avoiding the transfer process.
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Argon-40Ar, 99.95 atom %