Merck
  • Remote laser ablation electrospray ionization mass spectrometry for non-proximate analysis of biological tissues.
Remote laser ablation electrospray ionization mass spectrometry for non-proximate analysis of biological tissues.
Rapid communications in mass spectrometry : RCM (2014-12-03)
Laine R Compton, Brent Reschke, Jordan Friend, Matthew Powell, Akos Vertes
ABSTRACT
We introduce remote laser ablation electrospray ionization (LAESI), a novel, non-proximate ambient sampling technique. Remote LAESI allows additional analytical instrumentation to be incorporated during sample analysis. This work demonstrates the utility of remote LAESI and, when combined with optical microscopy, allows for the microscopy-guided sampling of biological tissues. Rapid prototyping using a 3D printer was applied to produce various ablation chamber geometries. A focused 5 ns, 2.94 µm laser pulse kept at 10 Hz ablated the sample within the chamber, remote to the mass spectrometer inlet. Ablated particulates were carried through a transfer tube by N2 gas, delivered to the electrospray plume and ionized. A long-distance microscope was used to capture images of tissues before, during and after ablation. Optimized remote LAESI was found to have a 27% transport efficiency compared with conventional LAESI, sufficient for many applications. A comparable molecular coverage was obtained with remote LAESI for the analysis of plant tissue. Proof-of-principle experiments using a pansy flower and a maple leaf indicated the functionality of this approach for selecting domains of interest for analysis by optical microscopy and obtaining chemical information from those selected regions by remote LAESI-MS. Remote LAESI is an ambient non-proximate sampling technique, proven to detect metabolites in biological tissues. When combined with optical microscopy, remote LAESI allows for the simultaneous acquisition of morphological and chemical information. This technique has important implications for histology, where chemical information for specific locations within a tissue is critical.
Product Description
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Millipore
Bifido Selective Supplement B, suitable for microbiology
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%