Merck
  • Bioactive thermoresponsive polyblend nanofiber formulations for wound healing.
Bioactive thermoresponsive polyblend nanofiber formulations for wound healing.
Materials science & engineering. C, Materials for biological applications (2015-01-13)
Mahesh D Pawar, G V N Rathna, Shubhang Agrawal, Bhanudas S Kuchekar
ABSTRACT
The rationale of this work is to develop new bioactive thermoresponsive polyblend nanofiber formulations for wound healing (topical). Various polymer compositions of thermoresponsive, poly(N-isopropylacrylamide), egg albumen and poly(ε-caprolactone) blend solutions with and without a drug [gatifloxacin hydrochloride, Gati] were prepared. Non-woven nanofibers of various compositions were fabricated using an electrospinning technique. The morphology of the nanofibers was analyzed by an environmental scanning electron microscope. The morphology was influenced by the concentration of polymer, drug, and polymer blend composition. Fourier transform infrared spectroscopy analysis showed the shift in bands due to hydrogen ion interactions between polymers and drug. Thermogram of PNIPAM/PCL/EA with Gati recorded a shift in lower critical solution temperature (LCST) and glass transition temperature (Tg) of PNIPAM. Similarly Tg and melting temperature (Tm) of PCL were shifted. X-ray diffraction patterns recorded a decrease in the crystalline state of PCL nanofibers and transformed crystalline drug to an amorphous state. In vitro release study of nanofibers with Gati showed initial rapid release up to 10h, followed by slow and controlled release for 696h (29days). Nanofiber mats with Gati exhibited antibacterial properties to Staphylococcus aureus, supported suitable controlled drug release with in vitro cell viability and in vivo wound healing.
Product Description
USP
1-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
N-Isopropylacrylamide, ≥99%
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%
Supelco
1-Propanol, analytical standard
Sigma-Aldrich
1-Propanol, ≥99%, FG
Supelco
1-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
Thiazole, 99%
Sigma-Aldrich
N-Isopropylacrylamide, 97%
Sigma-Aldrich
Thiazole, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®