Merck
  • Gambogic amide selectively upregulates TrkA expression and triggers its activation.
Gambogic amide selectively upregulates TrkA expression and triggers its activation.
Pharmacological reports : PR (2015-02-26)
Jianying Shen, Qingsheng Yu
ABSTRACT
Gambogic amide is the first identified small molecular agonist for TrkA receptor. It mimics NGF functions by selectively activating TrkA receptor and preventing neuron death. However, its function different from that of NGF remains unknown. In the current study, we detect the effect of gambogic amide on TrkA expression using TrkA-expressing cell lines in vitro and hippocampi from mice treated with gambogic amide. We have confirmed that gambogic amide displays robust neurotrophic activities in provoking neurite outgrowth in vitro. However, gambiogic amide displays a different kinetics from NGF in activating TrkA signals. NGF swiftly provokes TrkA activation and quickly induces TrkA degradation, while gambogic amid selectively upregulates TrkA protein and mRNA levels in a time-dependent manner. Administration of this compound in mice also activates TrkA receptor in hippocampus and promotes TrkA transcription and expression. This study provides a novel mechanism of how gambogic amide regulates TrkA receptor, other than mimicking NGF in triggering TrkA activation.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Supelco
L-Lysine, analytical standard
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder