Merck
  • Liquid chromatography-tandem mass spectrometry assay for the simultaneous quantification of simvastatin, lovastatin, atorvastatin, and their major metabolites in human plasma.
Liquid chromatography-tandem mass spectrometry assay for the simultaneous quantification of simvastatin, lovastatin, atorvastatin, and their major metabolites in human plasma.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-01-24)
Jiang Wang, Jasmine A Luzum, Mitch A Phelps, Joseph P Kitzmiller
ABSTRACT
Millions of individuals are treated with a variety of statins that are metabolized to a variety of active metabolites. A single assay capable of simultaneously quantifying commonly used statins and their major metabolites has not been previously reported. Herein we describe the development and validation of a novel and robust liquid chromatography-tandem mass spectrometry assay for simultaneously quantifying simvastatin, lovastatin, atorvastatin, and their metabolites, simvastatin acid, lovastatin acid, para-hydroxy atorvastatin, and ortho-hydroxy atorvastatin in human plasma. Plasma samples were processed with a simple protein precipitation technique using acetonitrile, followed by chromatographic separation using an Agilent Zorbax Extend C18 column. A 12.0min linear gradient elution was used at a flow rate of 400μL/min with a mobile phase of water and methanol, both modified with 2mM ammonium formate and 0.2% formic acid. The analytes and internal standard, hesperetin, were detected using the selected reaction monitoring mode on a TSQ Quantum Discovery mass spectrometer with positive electrospray ionization. The assay exhibited a linear range of 1-1000nM for simvastatin acid and lovastatin acid, and a linear range of 0.1-100nM for the other analytes in human plasma. The accuracy and the within- and between-day precisions of the assay were within acceptable ranges, and the method was successfully utilized to quantify the statins and their metabolites in human plasma samples collected from an ongoing pharmacokinetic study.
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Hesperetin, ≥95%
Supelco
Hesperetin, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%