Merck
  • Characterization of sakA gene from pathogenic dimorphic fungus Penicillium marneffei.
Characterization of sakA gene from pathogenic dimorphic fungus Penicillium marneffei.
International journal of medical microbiology : IJMM (2014-12-04)
Panjaphorn Nimmanee, Patrick C Y Woo, Aksarakorn Kummasook, Nongnuch Vanittanakom
ABSTRACT
Eukaryotes utilize stress activated protein kinase (SAPK) pathways to adapt to environmental stress, including heat, osmotic, oxidative or nutrient stresses. Penicillium marneffei (Talaromyces marneffei), the dimorphic pathogenic fungus that can cause disseminated mycosis in HIV-infected patients, has to encounter various types of stresses both outside and inside host cells. However, the strategies used by this fungus in response to these stresses are still unclear. In this report, the stress-activated kinase (sakA) gene of P. marneffei was characterized and the roles of this gene on various stress conditions were studied. The sakA gene deletion mutant was constructed using the split marker method. The phenotypes and sensitivities to varieties of stresses, including osmotic, oxidative, heat and cell wall stresses of the deletion mutant were compared with the wild type and the sakA complemented strains. Results demonstrated that the P. marneffei sakA gene encoded a putative protein containing TXY phosphorylation lip found in the stress high osmolarity glycerol 1 (Hog1)/Spc1/p38 MAPK family, and that this gene was involved not only in tolerance against oxidative and heat stresses, but also played a role in asexual development, chitin deposition, yeast cell generation in vitro and survival inside mouse and human macrophages.
Product Description
Trolamine impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Supelco
Aucubin, analytical standard
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
3-tert-Butyl-4-hydroxyanisole, ≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)
Sigma-Aldrich
Ethanolamine, ≥99%
Aucubin, primary reference standard
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Supelco
3-tert-Butyl-4-hydroxyanisole, Pharmaceutical Secondary Standard; Certified Reference Material