Merck
  • Serum myeloperoxidase/paraoxonase 1 ratio as potential indicator of dysfunctional high-density lipoprotein and risk stratification in coronary artery disease.
Serum myeloperoxidase/paraoxonase 1 ratio as potential indicator of dysfunctional high-density lipoprotein and risk stratification in coronary artery disease.
Atherosclerosis (2014-04-08)
Yoko Haraguchi, Ryuji Toh, Minoru Hasokawa, Hideto Nakajima, Tomoyuki Honjo, Kazunori Otsui, Kenta Mori, Maki Miyamoto-Sasaki, Masakazu Shinohara, Kunihiro Nishimura, Tatsuro Ishida, Ken-Ichi Hirata
ABSTRACT
Granular leukocyte-derived myeloperoxidase (MPO) promotes oxidation of lipoproteins, while paraoxonase 1 (PON1) has antioxidant properties for high-density lipoprotein (HDL). We evaluated their effects on coronary risk stratification and function of lipoproteins. A total 158 patients who had previously undergone percutaneous coronary intervention and who had been hospitalized for coronary re-angiography were enrolled. Coronary lesions (restenosis or de novo lesion) were observed in 84 patients but not associated with conventional lipid profile. In contrast, serum MPO levels and PON1 activities were significantly associated with the prevalence of coronary lesions. The high MPO/PON1 ratio, when cutoff values were set at 1.59, was independently correlated with restenosis (odds ratio 6.4, 95% CI 2.2-19.3, P = 0.001) and de novo lesions (odds ratio 3.5, 95% CI 1.3-9.4, P = 0.014). We isolated HDL from patients with high or low MPO/PON1 ratio, and compared anti-inflammatory properties of HDL. Human umbilical vein endothelial cells were stimulated with inflammatory cytokine, and the expression of vascular cell adhesion molecule-1 (VCAM-1) was evaluated. HDL isolated from patients with low serum MPO/PON1 ratio inhibited VCAM-1 expression significantly greater than that with high MPO/PON1 ratio. We also demonstrated that the cholesterol efflux capacity of apolipoprotein B-depleted serum from patients with high MPO/PON1 ratio was significantly decreased than that with low MPO/PON1 ratio. MPO/PON1 ratio could be a useful marker for secondary prevention of coronary artery disease through modulation of HDL function.
Product Description
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Phenyl acetate, 99%