Merck
  • Stability of 3-bromotyrosine in serum and serum 3-bromotyrosine concentrations in dogs with gastrointestinal diseases.
Stability of 3-bromotyrosine in serum and serum 3-bromotyrosine concentrations in dogs with gastrointestinal diseases.
BMC veterinary research (2015-01-18)
Panpicha Sattasathuchana, Niels Grützner, Rosana Lopes, Blake C Guard, Jan S Suchodolski, Jörg M Steiner
ABSTRACT
3-Bromotyrosine (3-BrY) is a stable product of eosinophil peroxidase and may serve as a marker of eosinophil activation. A gas chromatography/mass spectrometry method to measure 3-BrY concentrations in serum from dogs has recently been established and analytically validated. The aims of this study were to determine the stability of 3-BrY in serum, to determine the association between peripheral eosinophil counts and the presence of an eosinophilic infiltrate in the gastrointestinal tract, and to compare serum 3-BrY concentrations in healthy dogs (n = 52) and dogs with eosinophilic gastroenteritis (EGE; n = 27), lymphocytic-plasmacytic enteritis (LPE; n = 25), exocrine pancreatic insufficiency (EPI; n = 26), or pancreatitis (n = 27). Serum 3-BrY concentrations were stable for up to 8, 30, and 180 days at 4°C, -20°C, and -80°C, respectively. There was no significant association between peripheral eosinophil count and the presence of eosinophils in the GI tissues (P = 0.1733). Serum 3-BrY concentrations were significantly higher in dogs with EGE (median [range] = 5.04 [≤0.63-26.26] μmol/L), LPE (median [range] = 3.60 [≤0.63-15.67] μmol/L), and pancreatitis (median [range] = 1.49 [≤0.63-4.46] μmol/L) than in healthy control dogs (median [range] = ≤0.63 [≤0.63-1.79] μmol/L; P < 0.0001), whereas concentrations in dogs with EPI (median [range] = 0.73 [≤0.63-4.59] μmol/L) were not different compared to healthy control dogs. The present study revealed that 3-BrY concentrations were stable in serum when refrigerated and frozen. No relationship between peripheral eosinophil count and the presence of eosinophils infiltration in the GI tissues was found in this study. In addition, serum 3-BrY concentrations were increased in dogs with EGE, but also in dogs with LPE and pancreatitis. Further studies are needed to determine whether measurement of 3-BrY concentrations in serum may be useful to assess patients with suspected or confirmed EGE or LPE.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Supelco
Acetonitrile, analytical standard
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Trifluoroacetamide, 97%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
N,N-Diisopropylethylamine, 99.5%, biotech. grade
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
N-Ethyldiisopropylamine, ≥98.0%
Sigma-Aldrich
Undecane, ≥99%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Supelco
Undecane, analytical standard
Sigma-Aldrich
Undecane, ≥99%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material