Merck
  • Development and validation of an LC-MS/MS method for the determination of pelubiprofen and its active metabolite, trans-alcohol, in human plasma and its application to pharmacokinetic study.
Development and validation of an LC-MS/MS method for the determination of pelubiprofen and its active metabolite, trans-alcohol, in human plasma and its application to pharmacokinetic study.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-01-27)
Ju-Hee Ryu, Ji-Sun Park, Min-Ho Jo, Joo-Il Kim, Wang-Seob Shim, Bo-Hyung Kim, Sung-Vin Yim, Jongki Hong, Kyung-Tae Lee
ABSTRACT
A suitable liquid chromatography tandem mass spectrometry (LC-MS/MS) method is required to determine pelubiprofen and its active metabolite, trans-alcohol (M-D), in human plasma for pharmacokinetic studies of pelubiprofen preparations. After one-step liquid-liquid extraction (LLE) using methyl tert-butyl ether (MTBE), pelubiprofen, M-D, and tolbutamide (the internal standard, IS) were eluted from a Capcellpak C18 ACR column using a gradient mobile phase consisting of 0.1% formic acid in water and acetonitrile at a flow rate 0.35mL/min. The achieved lower limits of quantitation (LLOQ) of pelubiprofen and M-D were both 15ng/mL (S/N>10) and the standard calibration curves for pelubiprofen and M-D were linear (correlation coefficients >0.99) over the studied concentration range (15-2000ng/mL). Intra- and inter-day precisions were within 7.62% for all analytes and the deviation of assay accuracies was within ±13.23%. The developed method was successfully applied to a pharmacokinetic study of pelubiprofen in healthy Korean male volunteers.
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Supelco
Tolbutamide, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Tolbutamide, European Pharmacopoeia (EP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium formate, reagent grade, 97%