Merck
  • The urinary ratio of 3-hydroxykynurenine/3-hydroxyanthranilic acid is an index to predicting the adverse effects of D-tryptophan in rats.
The urinary ratio of 3-hydroxykynurenine/3-hydroxyanthranilic acid is an index to predicting the adverse effects of D-tryptophan in rats.
Journal of nutritional science and vitaminology (2014-10-10)
Katsumi Shibata, Tomoaki Ohno, Mitsue Sano, Tsutomu Fukuwatari
ABSTRACT
The adverse effects of D-tryptophan and the possibility of it being a surrogate index for predicting adverse effects in rats were investigated. Male rats were fed one of several test diets (20% casein diets with 0% (control), 0.1%, 0.2%, 0.3%, 0.4%, and 0.5% D-tryptophan) for 21 d, and 24-h urine samples on the final day of the experiment were collected. Analyses of food intake and body-weight changes revealed adverse effects to be observed in the group fed the 0.3% D-tryptophan diet. We propose urinary levels of 3-hydroxykynurenine/3-hydroxyanthranilic acid to be surrogate indicators for predicting the adverse effects of D-tryptophan from the break point of body-weight gains and urinary levels of D-tryptophan metabolites. The reaction 3-hydroxykynurenine→3-hydroxyanthranilic acid is catalyzed by the pyridoxal phosphate-dependent enzyme kynureninase. Increasing urinary 3-hydrokykynurenine indicates kynureninase deficiency. Intake of D-tryptophan in rats fed the 0.3% D-tryptophan diet was 0.21 g/kg body weight and feeding of the 0.3% D-tryptophan diet did not elicit adverse effects. Thus, the safe level of D-tryptophan was less than 0.2% in the diet, 0.15 g/kg body weight, in rats.
Product Description
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
2,3-Pyridinedicarboxylic acid, 99%
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
SAFC
L-Methionine
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
5-Hydroxyindole-3-acetic acid, ≥98% (HPLC), crystalline
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
L-Kynurenine, ≥98% (HPLC)
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard