Merck
  • Heterocyclic ring extension of estrone: synthesis and cytotoxicity of fused pyran, pyrimidine and thiazole derivatives.
Heterocyclic ring extension of estrone: synthesis and cytotoxicity of fused pyran, pyrimidine and thiazole derivatives.
Steroids (2014-04-02)
Rafat M Mohareb, Fatima Al-Omran, Rasha A Azzam
ABSTRACT
The one pot reaction of estrone with the aromatic aldehydes 2a-c and either of malononitrile or ethyl cyanoacetate afforded the fused pyran derivatives 4a-f. On the other hand, carrying the same reaction using thiourea instead of the cyanomethylene reagent gave the fused pyrimidine derivatives 6a-c. The latter compounds reacted with phenacyl bromide to give the thiazolo[3,2-a]pyrimidine derivatives 8a-c. The reaction of the title compound with bromine gave the monobromo derivative 13 which in turn reacted with either thiourea or cyanothioacetamide to give the thiazole derivatives 14 and 16, respectively. The cytotoxicity of the newly synthesized products was evaluated against six human cancer and normal cell lines where the results showed that compounds 4c, 4f, 6b, 8b, 8c, 10, 13, 16, 18c and 19c exhibited optimal cytotoxic effect against the cancer cell lines, with IC50's in the nM range.
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%