Merck
  • Mass spectrometric quantification of glucosylsphingosine in plasma and urine of type 1 Gaucher patients using an isotope standard.
Mass spectrometric quantification of glucosylsphingosine in plasma and urine of type 1 Gaucher patients using an isotope standard.
Blood cells, molecules & diseases (2015-04-07)
Mina Mirzaian, Patrick Wisse, Maria J Ferraz, Henrik Gold, Wilma E Donker-Koopman, Marri Verhoek, Herman S Overkleeft, Rolf G Boot, Gertjan Kramer, Nick Dekker, Johannes M F G Aerts
ABSTRACT
Deficiency of glucocerebrosidase (GBA) leads to Gaucher disease (GD), an inherited disorder characterised by storage of glucosylceramide (GlcCer) in lysosomes of tissue macrophages. Recently, we reported marked increases of deacylated GlcCer, named glucosylsphingosine (GlcSph), in plasma of GD patients. To improve quantification, [5-9] (13)C5-GlcSph was synthesised for use as internal standard with quantitative LC-ESI-MS/MS. The method was validated using plasma of 55 GD patients and 20 controls. Intra-assay variation was 1.8% and inter-assay variation was 4.9% for GlcSph (m/z 462.3). Plasma GlcSph levels with the old and new methods closely correlate (r=0.968, slope=1.038). Next, we analysed GlcSph in 24h urine samples of 30 GD patients prior to therapy. GlcSph was detected in the patient samples (median 1.20nM, range 0.11-8.92nM), but was below the limit of quantification in normal urine. Enzyme replacement therapy led to a decrease of urinary GlcSph of GD patients, coinciding with reductions in plasma GlcSph and markers of Gaucher cells (chitotriosidase and CCL18). In analogy to globotriaosylsphingsone in urine of Fabry disease patients, additional isoforms of GlcSph differing in structure of the sphingosine moiety were identified in GD urine samples. In conclusion, GlcSph can be sensitively detected by LC-ESI-MS/MS with an internal isotope standard. Abnormalities in urinary GlcSph are a hallmark of Gaucher disease allowing biochemical confirmation of diagnosis.
Sigma-Aldrich
D-Sphingosine, synthetic
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, 99.9%
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material