Merck
  • Neuronal marker recovery after Simvastatin treatment in dementia in the rat brain: in vivo magnetic resonance study.
Neuronal marker recovery after Simvastatin treatment in dementia in the rat brain: in vivo magnetic resonance study.
Behavioural brain research (2015-02-24)
Radka Tušková, Boris Lipták, Pavol Szomolányi, Oľga Vančová, Oľga Uličná, Zuzana Sumbalová, Jarmila Kucharská, Michal Dubovický, Siegfried Trattnig, Tibor Liptaj, Svatava Kašparová
ABSTRACT
The aim of study was to search for new biomarkers with a magnetic resonance technique to identify the early stages of dementia, induced by D-galactose, and evaluate Simvastatin therapy. Localized proton magnetic resonance spectroscopy measurements showed a significant decrease in the concentration of N-acetylaspartate+N-acetylaspartylglutamate and myo-inositol in the D-galactose group compared to the control group, and, conversely, an increase of N-acetylaspartate+N-acetylaspartylglutamate in the D-galactose/Simvastatin group. Using a saturation transfer experiment, with phosphorus magnetic resonance spectroscopy, we observed a significant elevation of the forward rate constant of the creatine kinase reaction in the brains of the D-galactose group compared to controls, and subsequently, a significant reduction of this reaction in the D-galactose/Simvastatin group. Spatial learning and memory were evaluated using the modified Morris water maze test. The dynamics of the learning process represented by the learning index revealed a significant reduction in learning in the D-galactose group, but the deficits as a consequence of the D-galactose effects were recovered in the D-galactose/Simvastatin group, in which the learning dynamics resembled those of the control group. By determining the thiobarbituric acid reactive substances and total coenzyme Q9 in plasma, we have shown that long-term administration of D-galactose created conditions for oxidative stress, and that the administration of Simvastatin decreased oxidative stress in plasma. Volumetry analyses from the hippocampal area show a reduction in the segmented area in the D-galactose group, compared with the control group, and an enlarged area in the hippocampus in the d-galactose/Simvastatin group.
Product Description
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Galactose, European Pharmacopoeia (EP) Reference Standard
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Galactose, United States Pharmacopeia (USP) Reference Standard