Merck
  • A rapid and sensitive method for hydroxyl radical detection on a microfluidic chip using an N-doped porous carbon nanofiber modified pencil graphite electrode.
A rapid and sensitive method for hydroxyl radical detection on a microfluidic chip using an N-doped porous carbon nanofiber modified pencil graphite electrode.
The Analyst (2014-05-20)
Jun Ouyang, Zhong-Qiu Li, Jing Zhang, Chen Wang, Jiong Wang, Xing-Hua Xia, Guo-Jun Zhou
ABSTRACT
Hydroxyl radicals (˙OH) play an important role in human diseases. Traditional detection methods are time consuming and require expensive instruments. Here, we present a simple and sensitive method for the detection of hydroxyl radicals on a microfluidic chip using an electrochemical technique. Aniline monomer is electrochemically polymerized on the surface of a pencil graphite electrode and carbonized at 800 °C. The resulting N-doped porous carbon nanofiber-modified pencil graphite electrode is embedded into a microfluidic chip directly as a working electrode. 4-Hydroxybenzoic acid (4-HBA) is selected as the trapping agent owing to its unique 3,4-DHBA product and high trapping efficiency. A low detection limit of 1.0 × 10(-6) M is achieved on the microfluidic chip. As a demonstration, the microfluidic chip is successfully utilized for the detection of ˙OH in cigarette smoke. The strong π-π stacking and hydrophobic interactions between the nitrogen-doped carbon materials and the pencil graphite make the modified electrode well-suited for the microfluidic chip.
Product Description
Supelco
3,4-Dihydroxybenzoic acid, analytical standard
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Supelco
4-Hydroxybenzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Protocatechuic acid, primary reference standard
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Hydroxyterephthalic acid, 97%
Sigma-Aldrich
Terephthalic acid, 98%
Supelco
Terephthalic acid, analytical standard