Merck
  • Exogenous heat shock protein gp96 ameliorates CD4+CD62L+ T-cell-mediated transfer colitis.
Exogenous heat shock protein gp96 ameliorates CD4+CD62L+ T-cell-mediated transfer colitis.
Inflammatory bowel diseases (2014-09-05)
Anne Fischbeck, Katja Schreiter, Katharina Leucht, Isabelle Frey-Wagner, Silvia Lang, Martin Hausmann, Michael Fried, Werner Falk, Gerhard Rogler
ABSTRACT
The heat shock protein gp96 is an endoplasmic reticulum chaperone involved in endoplasmic reticulum stress reactions. gp96 binds antigens and is secreted into extracellular space on cell stress. After reinternalization by antigen presenting cells, antigens can be transferred to major histocompatibility complex molecules. In recent studies, we found induction of gp96 during differentiation of intestinal macrophages, whereas it was absent in intestinal macrophages of patients with Crohn's disease. To study immuno-modulating effects of gp96 in T-cell transfer colitis BALB/c donor mice were injected with 2 × 100 μg gp96. After 1 week, 2.5 × 10(5) CD4+CD62L+ cells were isolated from spleens and injected into severe combined immunodeficiency recipients. Another group received cells from untreated donors and was treated with 100 μg gp96 after transfer. Control groups received cells from untreated donors, or buffer alone. After transfer of CD4+CD62L+ T cells from gp96-pretreated donors, mice (TBT gp96) showed an initial weight loss, but after 3 weeks, they recovered and reached the starting weight after 5 weeks. Mice treated with gp96 after transfer (TAT gp96) showed a delayed weight loss in comparison with the CD4+CD62L+ group. The histological scores in CD4CD62L mice were 2.6 ± 0.1, in TBT gp96 mice 1.3 ± 0.3 (CD4+CD62L+ versus TBT gp96: P < 0.05) and in mice treated after transfer 1.9 ± 0.1 (CD4+CD62L+ versus TAT gp96: P < 0.05). These findings indicate an essential role of gp96 in the maintenance of tolerance against luminal antigens in the intestinal mucosa. The absence of gp96 in intestinal macrophages of patients with Crohn's disease might provoke loss of this tolerance mediating mechanism.
Product Description
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%