Merck
  • Hyaluronic acid conjugated β-cyclodextrin-oligoethylenimine star polymer for CD44-targeted gene delivery.
Hyaluronic acid conjugated β-cyclodextrin-oligoethylenimine star polymer for CD44-targeted gene delivery.
International journal of pharmaceutics (2015-02-15)
Hui Yin, Feng Zhao, Daohai Zhang, Jun Li
ABSTRACT
A new CD44-targeted gene delivery system, the star-shaped cationic polymer containing a β-cyclodextrin (β-CD) core and multiple branched oligoethylenimine (OEI) arms with conjugated oligomer of hyaluronic acid (HA), was synthesized by reductive amination between β-CD-OEI star polymer and HA, and was characterized for pDNA condensation and nanoparticle formation, followed by evaluation for targeted gene delivery of luciferase reporter gene and wild type p53 gene in CD44-positive and CD44-negative cell lines. The β-CD-OEI-HA polymer contained 6 arms of OEI (600 Da) and a short HA segment. It could fully condense pDNA to form nanoparticles with sizes ranging from 100 to 200 nm at N/P ratios of 8 or higher. The conjugation of HA reduced cytotoxicity of β-CD-OEI-HA/pDNA polyplexes. It was found that CD44 receptor was highly expressed and localized at the membrane of MDA-MB-231 breast cancer cell line, while no CD44 was found at the membrane of MCF-7 epithelial cell line. Compared with PEI (25 kDa) and β-CD-OEI star polymers, β-CD-OEI-HA demonstrated significant increased gene transfection efficiency in MDA-MB-231 cells, while such effect was absent in MCF-7 cells. The targeted delivery of wild type p53 gene by β-CD-OEI-HA in MDA-MB-231 cells resulted in an increased cell cycle arrest at sub-G1 phase.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%