Merck
  • Imaging of mitochondrial Ca2+ dynamics in astrocytes using cell-specific mitochondria-targeted GCaMP5G/6s: mitochondrial Ca2+ uptake and cytosolic Ca2+ availability via the endoplasmic reticulum store.
Imaging of mitochondrial Ca2+ dynamics in astrocytes using cell-specific mitochondria-targeted GCaMP5G/6s: mitochondrial Ca2+ uptake and cytosolic Ca2+ availability via the endoplasmic reticulum store.
Cell calcium (2014-12-03)
Hailong Li, Xiaowan Wang, Nannan Zhang, Manoj K Gottipati, Vladimir Parpura, Shinghua Ding
ABSTRACT
Mitochondrial Ca(2+) plays a critical physiological role in cellular energy metabolism and signaling, and its overload contributes to various pathological conditions including neuronal apoptotic death in neurological diseases. Live cell mitochondrial Ca(2+) imaging is an important approach to understand mitochondrial Ca(2+) dynamics. Recently developed GCaMP genetically-encoded Ca(2+) indicators provide unique opportunity for high sensitivity/resolution and cell type-specific mitochondrial Ca(2+) imaging. In the current study, we implemented cell-specific mitochondrial targeting of GCaMP5G/6s (mito-GCaMP5G/6s) and used two-photon microscopy to image astrocytic and neuronal mitochondrial Ca(2+) dynamics in culture, revealing Ca(2+) uptake mechanism by these organelles in response to cell stimulation. Using these mitochondrial Ca(2+) indicators, our results show that mitochondrial Ca(2+) uptake in individual mitochondria in cultured astrocytes and neurons can be seen after stimulations by ATP and glutamate, respectively. We further studied the dependence of mitochondrial Ca(2+) dynamics on cytosolic Ca(2+) changes following ATP stimulation in cultured astrocytes by simultaneously imaging mitochondrial and cytosolic Ca(2+) increase using mito-GCaMP5G and a synthetic organic Ca(2+) indicator, x-Rhod-1, respectively. Combined with molecular intervention in Ca(2+) signaling pathway, our results demonstrated that the mitochondrial Ca(2+) uptake is tightly coupled with inositol 1,4,5-trisphosphate receptor-mediated Ca(2+) release from the endoplasmic reticulum and the activation of G protein-coupled receptors. The current study provides a novel approach to image mitochondrial Ca(2+) dynamics as well as Ca(2+) interplay between the endoplasmic reticulum and mitochondria, which is relevant for neuronal and astrocytic functions in health and disease.
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
SAFC
L-Cysteine
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland