Merck
  • Determination of N-nitrosodiethanolamine, NDELA in cosmetic ingredients and products by mixed mode solid phase extraction and UPLC-tandem mass spectrometry with porous graphitic carbon column through systemic sample pre-cleanup procedure.
Determination of N-nitrosodiethanolamine, NDELA in cosmetic ingredients and products by mixed mode solid phase extraction and UPLC-tandem mass spectrometry with porous graphitic carbon column through systemic sample pre-cleanup procedure.
Talanta (2015-03-17)
Kyung-Mi Joo, Mi-Sook Shin, Ji-hee Jung, Boo-Min Kim, John-Whan Lee, Hye-Jin Jeong, Kyung-Min Lim
ABSTRACT
A rapid, sensitive, accurate and specific ultra-performance liquid chromatography coupled to tandem mass spectrometry (UPLC-MS/MS) method for the detection of N-nitrosodiethanolamine (NDELA), a highly toxic contaminant in cosmetic raw materials and products was developed and validated. Systematized sample preparation steps were developed according to product types. Various SPE cartridges and columns were examined to establish the condition of SPE and chromatographic separation for NDELA. Sample cleanup steps consisting of solvent and liquid-liquid extraction tailored to the various sample matrix types were established prior to mixed mode SPE (Bond Elut AccuCAT). Chromatographic separation was achieved within 7 min on a porous graphitic carbon (PGC) column using a gradient elution with the mobile phase of 1mM ammonium acetate containing 0.1% acetic acid and methanol. NDELA was monitored using an electrospray positive ionization mass spectrometry in the multiple reaction monitoring (MRM) mode (m/z 134.9>103.7(quantifier) and 73.7(qualifier ion)) with d8-NDELA (m/z 143.1>111.0) as internal standard. The standard curves were linear over the concentration range of 1-100 ng/mL with a correlation coefficient higher than 0.99. The limit of detection (LOD) and the limit of quantification (LOQ) was 10 and 20 μg/kg, respectively (0.5 and 1 ng/mL in standard solution). The intra- and inter-day precisions were estimated to be below 11.1% and accuracies were within the range of 90.8-115.8%. The validated method was successfully applied to the analysis of real samples including raw materials, skin care, make-up, shampoos and hair products.
Product Description
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Nitrosodiethanolamine
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%