Merck
  • Impact on creatinine renal clearance by the interplay of multiple renal transporters: a case study with INCB039110.
Impact on creatinine renal clearance by the interplay of multiple renal transporters: a case study with INCB039110.
Drug metabolism and disposition: the biological fate of chemicals (2015-01-22)
Yan Zhang, Mark S Warren, Xuexiang Zhang, Sharon Diamond, Bill Williams, Naresh Punwani, Jane Huang, Yong Huang, Swamy Yeleswaram
ABSTRACT
Serum creatinine is commonly used as a marker of renal function, but increases in serum creatinine might not represent changes in glomerular filtration rate (GFR). INCB039110 (2-(3-(4-(7H-pyrrolo[2,3-day]pyrimidin-4-yl)-1H-pyrazol-1-yl)-1-(1-(3-fluoro-2-(trifluoromethyl)isonicotinoyl)piperidin-4-yl)azetidin-3-yl)acetonitrile) is an inhibitor of the Janus kinases (JAKs) with selectivity for JAK1. In a phase 1 study, a modest and reversible increase in serum creatinine was observed after treatment with INCB039110. However, a dedicated renal function study with INCB039110, assessed by iohexol plasma clearance, conducted in healthy volunteers indicated no change in GFR. In vitro studies were therefore conducted to investigate the interaction of INCB039110 with five transporters that are likely involved in the renal clearance of creatinine. Cell systems expressing individual or multiple transporters were used, including a novel quintuple-transporter model OAT2/OCT2/OCT3/MATE1/MATE2-K. INCB039110 potently inhibited OCT2-mediated uptake of creatinine as well as MATE1-/MATE2-K-mediated efflux of creatinine. Given the interactions of INCB039110 with multiple transporters affecting creatinine uptake and efflux, an integrated system expressing all five transporters was sought; in that system, INCB039110 caused a dose-dependent decrease in transcellular transport of creatinine with weaker net inhibition compared with the effects on individual transporters. In summary, a molecular mechanism for the increase in serum creatinine by INCB039110 has been established. These studies also underline the limitations of using serum creatinine as a marker of renal function.
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Histodenz, nonionic density gradient medium
Supelco
Iohexol, analytical standard
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Iohexol, European Pharmacopoeia (EP) Reference Standard
Iohexol for peak identification, European Pharmacopoeia (EP) Reference Standard