Merck
  • A graphene/zinc oxide nanocomposite film protects dental implant surfaces against cariogenic Streptococcus mutans.
A graphene/zinc oxide nanocomposite film protects dental implant surfaces against cariogenic Streptococcus mutans.
Biofouling (2014-11-29)
Shatavari Kulshrestha, Shakir Khan, Ramovatar Meena, Braj R Singh, Asad U Khan
ABSTRACT
Oral biofilms play a crucial role in the development of dental caries and other periodontal diseases. Streptococcus mutans is one of the primary etiological agents in dental caries. Implant systems are regularly employed to replace missing teeth. Oral biofilms accumulate on these implants and are the chief cause of dental implant failure. In the present study, the potential of graphene/zinc oxide nanocomposite (GZNC) against the cariogenic properties of Streptococcus mutans was explored and the anti-biofilm behaviour of artificial acrylic teeth surfaces coated with GZNC was examined. Acrylic teeth are a good choice for implants as they are low cost, have low density and can resist fracture. Microscopic studies and anti-biofilm assays showed a significant reduction in biofilm in the presence GZNC. GZNC was also found to be nontoxic against HEK-293 (human embryonic kidney cell line). The results indicate the potential of GZNC as an effective coating agent for dental implants by efficiently inhibiting S. mutans biofilms.
Product Description
USP
Menadione, United States Pharmacopeia (USP) Reference Standard
Menadione, European Pharmacopoeia (EP) Reference Standard
Supelco
Menadione, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Menadione (K3), analytical standard
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Menadione, meets USP testing specifications
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use