Merck
  • Short-chain fatty acids and acidic pH upregulate UT-B, GPR41, and GPR4 in rumen epithelial cells of goats.
Short-chain fatty acids and acidic pH upregulate UT-B, GPR41, and GPR4 in rumen epithelial cells of goats.
American journal of physiology. Regulatory, integrative and comparative physiology (2014-12-19)
Zhongyan Lu, Hongbing Gui, Lei Yao, Lei Yan, Holger Martens, Jörg R Aschenbach, Zanming Shen
ABSTRACT
Currently, the mechanism(s) responsible for the regulation of urea transporter B (UT-B) expression levels in the epithelium of the rumen remain unclear. We hypothesized that rumen fermentation products affect ruminal UT-B expression. Therefore, the effects of short-chain fatty acids (SCFA), pH, ammonia, and urea on mRNA and protein levels of UT-B were assayed in primary rumen epithelial cell cultures and in rumen epithelium obtained from intact goats. In vitro, SCFA and acidic pH were found to synergetically stimulate both mRNA and protein expression of UT-B, whereas NH4Cl decreased mRNA and protein levels of UT-B at pH 6.8. Treatment with urea increased both levels at pH 7.4. When goats received a diet rich in nitrogen (N) and nonfiber carbohydrates (NFC), their rumen epithelium had higher levels of UT-B, and the rumen contained higher concentrations of SCFA and NH3-N with a lower pH. An increase in plasma urea-N concentration was also observed compared with the plasma of the goats that received a diet low in N and NFC. In a second feeding trial, goats that received a NFC-rich, but isonitrogenous, diet had higher mRNA and protein levels of UT-B, and higher levels of G protein-coupled receptor (GPR) 41 and GPR4, in their rumen epithelium. The ruminal concentrations of SCFA and NH3-N also increased, while a lower pH was detected. In contrast, the serum urea-N concentrations remained unchanged. These data indicate that ruminal SCFA and pH are key factors, via GPR4 and GPR41, in the dietary regulation of UT-B expression, and they have priority over changes in plasma urea.
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
Sodium butyrate, 98%
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Supelco
Sodium butyrate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
SAFC
L-Glutamine
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis