Merck
  • Pristane-induced granulocyte recruitment promotes phenotypic conversion of macrophages and protects against diffuse pulmonary hemorrhage in Mac-1 deficiency.
Pristane-induced granulocyte recruitment promotes phenotypic conversion of macrophages and protects against diffuse pulmonary hemorrhage in Mac-1 deficiency.
Journal of immunology (Baltimore, Md. : 1950) (2014-10-05)
Yiqin Shi, Naotake Tsuboi, Kazuhiro Furuhashi, Qiuna Du, Asuka Horinouchi, Kayaho Maeda, Tomoki Kosugi, Seiichi Matsuo, Shoichi Maruyama
ABSTRACT
Diffuse pulmonary hemorrhage (DPH) is an uncommon but critical complication of systemic lupus erythematosus. Peritoneal administration of 2,6,10,14-tetramethylpentadecane (pristane) can recapitulate a lupus-like syndrome in mice, which can develop into DPH within a few weeks, especially in C57BL/6 mice. Mac-1 (CD11b/CD18), a leukocyte adhesion molecule, is known to play a role in inflammation by regulating migration of leukocytes into injured tissue. In this study, we aimed to clarify the role of Mac-1 in pristane-induced DPH, using Mac-1(-/-) and wild-type (WT) mice on a C57BL/6 background. After pristane injection, Mac-1(-/-) mice showed reduced prevalence of DPH and attenuated peritonitis compared with WT mice. Analysis of the peritoneal lavage on days 5 and 10 after pristane treatment revealed increased numbers of eosinophils and alternatively activated macrophages, but decreased numbers of neutrophils and classically activated macrophages in Mac-1(-/-) mice compared with WT. Enhanced production of IL-4 and IL-13, both key mediators of macrophage polarization toward the mannose receptor(+) (MMR(+)) phenotype, was observed in the peritoneal cavity of Mac-1(-/-) mice. Depletion of neutrophils and eosinophils or adoptive transfer of classically activated macrophages resulted in the exacerbation of pristane-mediated DPH in both WT and Mac-1(-/-) mice. Moreover, peritoneal transfer of F4/80(high)MMR(+) alternatively activated macrophages successfully reduced the prevalence of DPH in WT mice. Collectively, Mac-1 promoted acute inflammatory responses in the peritoneal cavity and the lungs by downregulating granulocyte migration and subsequent phenotypic conversion of macrophages in a pristane-induced systemic lupus erythematosus model.
Product Description
SAFC
Pristane, synthetic, liquid, sterile-filtered, BioReagent