Merck
  • Memantine plus vitamin D prevents axonal degeneration caused by lysed blood.
Memantine plus vitamin D prevents axonal degeneration caused by lysed blood.
ACS chemical neuroscience (2015-01-15)
David Charier, Olivier Beauchet, Morgane Bell, Bernard Brugg, Robert Bartha, Cedric Annweiler
ABSTRACT
Intracranial hemorrhage, whether due to traumatic brain injury or ruptured cerebral aneurysm, is characterized by major neurological damage and a high mortality rate. Apart from cerebral vasospasm and mass effect, brain injury results from the release of unclotted blood that contacts neurons causing calcic stress. The combination of memantine with vitamin D, a neurosteroid hormone, may prevent blood neurotoxicity. Our purpose was to examine the potential protective effects of memantine + vitamin D against lysed or clotted blood in cortical neuronal cultures. We provide the first evidence that cortical axons in contact with lysed blood degenerate less after exposure to lysed blood in microfluidic neuronal cultures enriched with both memantine and vitamin D compared to control medium and cultures enriched with only memantine or only vitamin D. The reported synergistic neuroprotective effect of memantine + vitamin D, the combination originating an effect stronger than the sum, strongly encourages using both drugs following intracranial hemorrhage.
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM