Merck
  • Dissolvable layered double hydroxide coated magnetic nanoparticles for extraction followed by high performance liquid chromatography for the determination of phenolic acids in fruit juices.
Dissolvable layered double hydroxide coated magnetic nanoparticles for extraction followed by high performance liquid chromatography for the determination of phenolic acids in fruit juices.
Journal of chromatography. A (2014-09-28)
Mohammad Saraji, Milad Ghani
ABSTRACT
A magnesium-aluminum layered double hydroxide coated on magnetic nanoparticles was synthesized and used as a sorbent to extract some phenolic acids including p-hydroxy benzoic acid, caffeic acid, syringic acid, p-coumaric acid and ferulic acid from fruit juices. After extraction, the elution step was performed through dissolving double hydroxide layers containing the analytes by changing the solution pH. The extracted phenolic acids were separated and quantified using high performance liquid chromatography-photodiode array detection. Experimental parameters such as sorbent amount, solution pH, desorption solvent volume and extraction time were studied and optimized. The linearity range of the method was between 2 and 500μgL(-1) with the determination coefficient (r(2)) higher than 0.991. Relative standard deviations for intra- and inter-day precision for the analytes at 100μgL(-1) were in the range of 4.3-9.2% and 4.9-8.6%, respectively. Batch-to-batch reproducibility at 100μgL(-1) concentration level was in the range of 7.8-11% (n=3). The limits of detection were between 0.44 and 1.3μgL(-1). Relative recoveries higher than 81% with RSDs in the range of 4.2-9.7% were obtained in the analysis of fruit juice samples.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Methanol, analytical standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Iron(III) chloride hexahydrate, puriss. p.a., ACS reagent, crystallized, 98.0-102% (RT)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Iron(III) chloride hexahydrate, ACS reagent, 97%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Iron(III) chloride hexahydrate, reagent grade, ≥98%, chunks
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
trans-Ferulic acid, 99%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
trans-Ferulic acid, ≥99%