Merck
  • Circulating myeloid-derived suppressor cells correlate with clinical outcome in Hodgkin Lymphoma patients treated up-front with a risk-adapted strategy.
Circulating myeloid-derived suppressor cells correlate with clinical outcome in Hodgkin Lymphoma patients treated up-front with a risk-adapted strategy.
British journal of haematology (2014-11-08)
Alessandra Romano, Nunziatina Laura Parrinello, Calogero Vetro, Stefano Forte, Annalisa Chiarenza, Amalia Figuera, Giovanna Motta, Giuseppe Alberto Palumbo, Massimo Ippolito, Ugo Consoli, Francesco Di Raimondo
ABSTRACT
In the attempt to find a peripheral blood biological marker that could mirror the dysregulated microenvironment of Hodgkin Lymphoma (HL), we analysed the amount of myeloid-derived suppressor cells (MDSC), including the three main sub-types (monocytic, granulocytic and CD34 + fraction). The absolute MDSC count was investigated in 60 consecutive newly diagnosed HL patients and correlated with clinical variables at diagnosis and outcome. Patients received standard-of-care chemotherapy with the exception of interim fluorodeoxyglucose positron emission tomography (PET-2)-positive patients, who were switched early to a salvage regimen. All MDSC subsets were increased in HL patients compared to normal subjects (P < 0·0001) and were higher in non-responders. However, a strong prognostic significance was limited to immature (CD34(+) ) MDSC. A cut-off level of 0·0045 × 10(9) /l for CD34(+) MDSC resulted in 89% (95% confidence interval [CI] 52-99%) sensitivity and 92% (95% CI 81-98%) specificity. The positive predictive value to predict progression-free survival was 0·90 for PET-2 and 0·98 for CD34(+) MDSC count; the negative predictive value was 0·57 for PET-2 and 0·73 for CD34(+) MDSC. PFS was significantly shorter in patients with more than 0·0045 × 10(9) CD34(+) MDSC cells/l at diagnosis and/or PET-2 positivity (P < 0·0001). In conclusion, all circulating MDSC subsets are increased in HL; CD34(+) MDSC predict short PFS, similarly to PET-2 but with the advantage of being available at diagnosis.
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Ammonium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
O-Acetyl-L-carnitine hydrochloride, ≥99% (titration), powder
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)