Merck
  • Gaseous emissions in municipal wastes composting: effect of the bulking agent.
Gaseous emissions in municipal wastes composting: effect of the bulking agent.
Bioresource technology (2014-10-02)
Caterina Maulini-Duran, Adriana Artola, Xavier Font, Antoni Sánchez
ABSTRACT
In this study, the emissions of volatile organic compounds (VOC), CH4, N2O and NH3 during composting non-source selected MSW, source selected organic fraction of municipal solid wastes (OFMSW) with wood chips as bulking agent (OF_wood) and source selected OFMSW with polyethylene (PE) tube as bulking agent (OF_tube) and the effect of bulking agent on these emissions have been systematically studied. Emission factors are provided (in kg compound Mg(-1) dry matter): OF_tube (CH4: 0.0185±0.004; N2O: 0.0211±0.005; NH3: 0.612±0.269; VOC: 0.688±0.082) and MSW (CH4: 0.0549±0.0171; N2O: 0.032±0.015; NH3: 1.00±0.20; VOC: 1.05±0.18) present lower values than OF_wood (CH4: 1.27±0.09; N2O: 0.021±0.006; NH3: 4.34±2.79; VOC: 0.989±0.249). A detailed composition of VOC is also presented. Terpenes were the main emitted VOC family in all the wastes studied. Higher emissions of alpha and beta pinene were found during OF_wood composting processes.
Product Description
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Manual Holder, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, red hub
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, white hub
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Portable Field Sampler, coating PDMS