Merck
  • Comparison of five different C18 HPLC analytical columns for the analysis of ochratoxin A in different matrices.
Comparison of five different C18 HPLC analytical columns for the analysis of ochratoxin A in different matrices.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-10-02)
Y Sultan, N Magan, A Medina
ABSTRACT
The performance of five different C18 chromatography analytical columns with different lengths, particle sizes and porosities were compared for analysis of ochratoxin A (OTA) in fungal cultures and raisin samples. Chromatographic parameters including retention time, limit of detection, limit of quantification, number of theoretical plates and reduced plate height were obtained and compared. This showed that, compared with traditional columns, shorter ones (100 and 75mm×4.6mm) with 2.7μm solid core particles are suitable for analysis of OTA in different matrices and allows a reduction of the total analysis time by approximately 50% without any detrimental effect on performance. This leads to significant reduction in analysis costs by savings in use of organic solvents and increasing the total number of analyses per day. The capability of these columns for analyzing samples, from different matrices, was assessed by analyzing OTA-contaminated samples from cultures of Aspergillus westerdijkiae and Aspergillus niger grown on a defined nutritional media (yeast extract sucrose agar) and from natural and OTA spiked raisins.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard