Merck
  • Plasma DNA integrity as a biomarker for primary and metastatic breast cancer and potential marker for early diagnosis.
Plasma DNA integrity as a biomarker for primary and metastatic breast cancer and potential marker for early diagnosis.
Breast cancer research and treatment (2014-05-20)
Dharanija Madhavan, Markus Wallwiener, Karin Bents, Manuela Zucknick, Juliane Nees, Sarah Schott, Katarina Cuk, Sabine Riethdorf, Andreas Trumpp, Klaus Pantel, Christof Sohn, Andreas Schneeweiss, Harald Surowy, Barbara Burwinkel
ABSTRACT
Circulating or cell-free DNA (cfDNA) has been evaluated as a biomarker in many cancers including breast cancer. In particular, integrity of cfDNA has been shown to be altered in cancers. We have estimated the biomarker potential of cfDNA in primary (PBC) and metastatic breast cancer (MBC). cfDNA integrity (cfDI) and concentration were determined in plasma of 383 individuals, including 82 PBC and 201 MBC cases, as well as 100 healthy controls, by measuring ALU and LINE1 repetitive DNA elements using quantitative PCR. The MBC patient group was further sub-divided into patients with detectable circulating tumour cells (CTCpos-MBC, n = 100) and those without (CTCneg-MBC, n = 101). A hierarchical decrease in cfDI and increase in cfDNA concentration from healthy controls to PBC and further onto MBC patients were observed. Investigation of cfDNA in media of cell lines was in concordance with these results. Combination of cfDI and cfDNA concentration could differentiate PBC cases from controls (area under the curve, AUC = 0.75), MBC cases from controls (AUC = 0.81 for CTCneg-MBC, AUC = 0.93 for CTCpos-MBC), and CTCneg-MBC from CTCpos-MBC cases (AUC = 0.83). cfDI additionally demonstrated a positive correlation to progression-free (HR of 0.46 for ALU, P = 0.0025) and overall survival (HR of 0.15 for ALU and 0.20 for LINE1, P < 0.0001) in MBC, and had lower prediction error than CTC status. Our findings show that reduced cfDI and increased cfDNA concentration can serve as diagnostic markers for PBC and MBC, and cfDI as a prognostic marker for MBC, thereby making them attractive candidates for blood-based multi-marker assays.
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder