Merck
  • Quantification of leakage from large unilamellar lipid vesicles by fluorescence correlation spectroscopy.
Quantification of leakage from large unilamellar lipid vesicles by fluorescence correlation spectroscopy.
Biochimica et biophysica acta (2014-08-20)
Kasper Kristensen, Jonas R Henriksen, Thomas L Andresen
ABSTRACT
Fluorescence correlation spectroscopy (FCS) is a powerful experimental technique that in recent years has found numerous applications for studying biological phenomena. In this article, we scrutinize one of these applications, namely, FCS as a technique for studying leakage of fluorescent molecules from large unilamellar lipid vesicles. Specifically, we derive the mathematical framework required for using FCS to quantify leakage of fluorescent molecules from large unilamellar lipid vesicles, and we describe the appropriate methodology for successful completion of FCS experiments. By use of this methodology, we show that FCS can be used to accurately quantify leakage of fluorescent molecules from large unilamellar lipid vesicles, including leakage of fluorescent molecules of different sizes. To demonstrate the applicability of FCS, we have investigated the antimicrobial peptide mastoparan X. We show that mastoparan X forms transient transmembrane pores in POPC/POPG (3:1) vesicles, resulting in size-dependent leakage of molecules from the vesicles. We conclude the paper by discussing some of the advantages and limitations of FCS as compared to other existing methods to measure leakage from large unilamellar lipid vesicles.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Piperazine, ReagentPlus®, 99%
Sigma-Aldrich
Ethanesulfonic acid, 95%
Sigma-Aldrich
Piperazine, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
Rhodamine 6G, analytical standard
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent