Merck
  • Vitamin D₃ metabolites enhance the NLRP3-dependent secretion of IL-1β from human THP-1 monocytic cells.
Vitamin D₃ metabolites enhance the NLRP3-dependent secretion of IL-1β from human THP-1 monocytic cells.
Journal of cellular biochemistry (2015-02-03)
Sarah E Tulk, Kuo-Chieh Liao, Daniel A Muruve, Yan Li, Paul L Beck, Justin A MacDonald
ABSTRACT
Vitamin D3 has emerged as an important regulator of the immune system. With metabolic enzymes for vitamin D3 activation and vitamin D receptors (VDR) now identified in a variety of immune cells, the active vitamin D3 metabolite 1,25(OH)2D3, is thought to possess immunomodulatory properties. We examined whether 1,25(OH)2D3 might also enhance the NLRP3-dependent release of mature IL-1β from macrophages. PMA-differentiated THP-1 cells were stimulated with vitamin D3 metabolites and assessed for CYP27, CYP24, NLRP3, ASC, pro-caspase-1 expression by western blot and real-time qPCR as well as inflammasome activation with pro-inflammatory cytokine IL-1β release measured by ELISA. Exposure to 1,25(OH)2D3 had no effect on the basal expression levels of VDR; however, CYP27A1 transcript was suppressed and CYP24A1 transcript was substantively elevated. Both 1,25(OH)2D3 - and 25(OH)D3 induced IL-1β release from THP-1 cells, and these effects were blocked with application of the caspase-1 inhibitor YVAD and the NLRP3 inhibitors glyburide and Bay 11-7082. Interestingly, 1,25 (OH)2D3 exposure reduced NLRP3 protein expression but had no effect on ASC or pro-caspase-1 protein levels. The increase in mature IL-1β elicited by 1,25(OH)2D3 was modest compared to that found for ATP or C. difficile toxins. However, co-treatment of THP-1 cells with ATP and 1,25(OH)2D3 resulted in more IL-1β secretion than ATP or 1,25(OH)2D3 alone.
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Supelco
Cholecalciferol (D3), analytical standard
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Glyburide (Glibenclamide), Pharmaceutical Secondary Standard; Certified Reference Material
Glibenclamide for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Glybenclamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glyburide, meets USP testing specifications
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Glybenclamide, ≥99% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%