Merck
  • Triterpenoids of sour jujube show pronounced inhibitory effect on human tumor cells and antioxidant activity.
Triterpenoids of sour jujube show pronounced inhibitory effect on human tumor cells and antioxidant activity.
Fitoterapia (2014-08-05)
Aimin Qiao, Yihai Wang, Limin Xiang, Zhenxue Zhang, Xiangjiu He
ABSTRACT
Sour jujube is a common fruit and traditional medicine in China. Bioactivity-guided fractionation of sour jujube was used to determine the chemical identity of potent antiproliferative and antioxidant constituents. Four novel ursane-type triterpenoids, together with 8 known were isolated and identified. The new triterpenoids were elucidated to be 2α,3β,13β,23-tetrahydroxy-urs-11-en-28-oic acid (3), 2α,3β-dihydroxy-urs-20(30)-en-28-oic acid (9), 2α,3β,28-trihydroxy-urs-20(30)-ene (10), and 3β,12β,13β-trihydroxy-ursan-28-oic acid (11). Among the triterpenoids isolated, 2α,3β,19α-trihydroxy-urs-12-en-28-oic acid (7), 9 and 10 showed high potent inhibitory activity toward the proliferation of HepG2 cells, which the IC50 values were lower than 5 μM. Compounds 9 and 10 also exhibited pronounced activity against MCF-7 cells, with IC50 value of 0.8 ± 0.03 and 1.5 ± 0.1 μM, respectively. Compound 10 showed high antioxidant activity with an EC50 of 0.8 ± 0.02 μM, which was 18.9 times higher than ascorbic acid in antioxidant capacity.
Product Description
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, anhydrous, 95%