Merck
  • Mitragynine 'Kratom' related fatality: a case report with postmortem concentrations.
Mitragynine 'Kratom' related fatality: a case report with postmortem concentrations.
Journal of analytical toxicology (2014-12-18)
Iain M McIntyre, Amber Trochta, Susan Stolberg, Steven C Campman
ABSTRACT
A 24-year-old man whose medical history was significant for alcohol abuse and depression was found unresponsive in bed. He had several prior suicide attempts with 'pills' and had also been hospitalized for an accidental overdose on a previous occasion. Autopsy findings were unremarkable apart from pulmonary edema and congestion, and urinary retention. Postmortem peripheral blood initially screened positive for mitragynine 'Kratom' (by routine alkaline drug screen by gas chromatography-mass spectrometry, GC-MS), which was subsequently confirmed by a specific GC-MS selective ion mode analysis following solid-phase extraction. Concentrations were determined in the peripheral blood (0.23 mg/L), central blood (0.19 mg/L), liver (0.43 mg/kg), vitreous (<0.05 mg/L), urine (0.37 mg/L) and was not detected in the gastric. Therapeutic concentrations of venlafaxine, diphenhydramine and mirtazapine were also detected together with a negligible ethanol of 0.02% (w/v). The results are discussed in relation to previous cases of toxicity, and the lack of potential for mitragynine postmortem redistribution.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Supelco
Methanol, analytical standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%