Merck
  • Development and validation of liquid chromatography tandem mass spectrometry method for simultaneous quantification of first line tuberculosis drugs and metabolites in human plasma and its application in clinical study.
Development and validation of liquid chromatography tandem mass spectrometry method for simultaneous quantification of first line tuberculosis drugs and metabolites in human plasma and its application in clinical study.
Journal of pharmaceutical and biomedical analysis (2014-12-03)
Kim H Hee, Jerold J Seo, Lawrence S Lee
ABSTRACT
Rifampicin (RIF) and isoniazid (INH), first line drugs for the treatment of tuberculosis, are known to cause hepatotoxicity as a serious adverse side effect. To further understand the pharmacokinetic parameters of these two drugs, we have developed and validated a rapid, sensitive and selective LC-MS/MS method for simultaneous quantification of RIF, INH and their metabolites 25-desacetylrifampicin (DRIF), acetylisoniazid (AcINH) and isonicotinic acid (INA). Analytes were extracted from 20 μl of plasma using solid-phase extraction (SPE) followed by chromatographic separation on Zorbax SB-Aq column (50 mm × 4.6mm, particle size 5 μm) using stepwise gradient elution of 5mM ammonium acetate and 90% acetonitrile with 0.1% formic acid. Separation of all analytes was achieved in the total run time of 6 min. The analytes were detected under positive ionization mode by multiple reaction monitoring (MRM) and quantification of analytes was performed by using deuterium-labelled internal standard. Excellent linearity (r(2) ≥ 0.995) was achieved for the analytes at different concentration ranges. The method was accurate (90-115%), precise (CV %<14) and specific. Matrix effect was in the range of 93-111% except for INA (40-42%) while recovery from SPE was reproducible (CV %<7.4) in the range of 60-86%. Post-preparative stability (48 h, 6°C autosampler) and freeze-thaw stability (3 cycles) were assessed with mean recovered concentration of >85%. The method was successfully applied to a clinical study of 33 healthy subjects to evaluate the effect of concomitant of INH on the pharmacokinetic parameters of RIF as well as the segregation of the subjects into slow or fast acetylators of INH.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Isoniazid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Deuterium, 99.8 atom % D
Sigma-Aldrich
Deuterium, 99.9 atom % D
Sigma-Aldrich
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%