Merck
  • Atomoxetine reduces hyperactive/impulsive behaviours in neurokinin-1 receptor 'knockout' mice.
Atomoxetine reduces hyperactive/impulsive behaviours in neurokinin-1 receptor 'knockout' mice.
Pharmacology, biochemistry, and behavior (2014-12-03)
Katharine Pillidge, Ashley J Porter, Temis Vasili, David J Heal, S Clare Stanford
ABSTRACT
Mice with functional ablation of the neurokinin-1 receptor gene (NK1R(-/-)) display behavioural abnormalities which resemble the hyperactivity, inattention and impulsivity seen in Attention Deficit Hyperactivity Disorder (ADHD). Here, we investigated whether the established ADHD treatment, atomoxetine, alleviates these abnormalities when tested in the light/dark exploration box (LDEB) and 5-Choice Serial Reaction-Time Task (5-CSRTT). Separate cohorts of mice were tested in the 5-CSRTT and LDEB after treatment with no injection, vehicle or atomoxetine (5-CSRTT: 0.3, 3 or 10mg/kg; LDEB: 1, 3 or 10mg/kg). Atomoxetine reduced the hyperactivity displayed by NK1R(-/-) mice in the LDEB at a dose (3mg/kg) which did not affect the locomotor activity of wildtypes. Atomoxetine (10mg/kg) also reduced impulsivity in NK1R(-/-) mice, but not wildtypes, in the 5-CSRTT. No dose of drug affected attention in either genotype. This evidence that atomoxetine reduces hyperactive/impulsive behaviours in NK1R(-/-) mice consolidates the validity of using NK1R(-/-) mice in research of the aetiology and treatment of ADHD.
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether