Merck
  • A simple method for simultaneous determination of N-arachidonoylethanolamine, N-oleoylethanolamine, N-palmitoylethanolamine and 2-arachidonoylglycerol in human cells.
A simple method for simultaneous determination of N-arachidonoylethanolamine, N-oleoylethanolamine, N-palmitoylethanolamine and 2-arachidonoylglycerol in human cells.
Analytical and bioanalytical chemistry (2014-12-19)
Igor Ivanov, Philipp Borchert, Burkhard Hinz
ABSTRACT
The endocannabinoid system has been considered as a target for pharmacological intervention. Accordingly, inhibition of fatty acid amide hydrolase (FAAH), a degrading enzyme of the endocannabinoids N-arachidonoylethanolamine (anandamide; AEA) and 2-arachidonoylglycerol (2-AG) as well as of the endocannabinoid-like substances N-oleoylethanolamine (OEA) and N-palmitoylethanolamine (PEA), can cause augmented endogenous cannabinoid tone. Using liquid chromatography coupled with positive electrospray ionisation mass spectrometry, we herein describe a method to simultaneously quantify levels of AEA, OEA, PEA and 2-AG in cultured cells. The procedure was developed according to the FDA guidelines for bioanalytical methods validation. The limits of quantification (LOQs) were 0.05 pmol for AEA, 0.09 pmol for OEA, 0.10 pmol for PEA and 0.80 pmol for 2-AG when molecular ion monitoring was used. In H460 human lung carcinoma cells, basal levels of all four analytes ranged between 2 and 17 pmol mg(-1) protein with PEA showing the lowest and OEA the highest concentrations. Endocannabinoid levels observed in mesenchymal stem cells were of the same order of magnitude when compared to those in H460 human lung carcinoma cells.
Sigma-Aldrich
N-Oleoylethanolamine, ~98% (TLC)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Trizma® base, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Palmitoylethanolamide
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Arachidonylethanolamide, ≥97.0% (TLC), oil