Merck
  • Sperm distribution and fertilization after unilateral and bilateral laparoscopic artificial insemination with frozen-thawed goat semen.
Sperm distribution and fertilization after unilateral and bilateral laparoscopic artificial insemination with frozen-thawed goat semen.
Theriogenology (2014-09-02)
Nitira Anakkul, Junpen Suwimonteerabutr, Theerawat Tharasanit, Sarawanee Khunmanee, Paweena Diloksumpan, Debra K Berg, Mongkol Techakumphu
ABSTRACT
Generally, laparoscopic artificial insemination (LAI) provides a higher success rate than of cervical insemination in goats. However, the sperm distribution after LAI in goats remains unknown, particularly when frozen-thawed semen is used. This study evaluated the distribution of frozen-thawed goat spermatozoa after LAI and compared the effects of sperm numbers and deposition sites (unilateral and bilateral sites) on pregnancy rate. In experiment 1, the frozen-thawed spermatozoa were stained either with CellTracker Green CMFDA (CT-Green) or CellTracker Red CMPTX (CT-Red), and in vitro evaluations of viability and motility were performed. In experiment 2, the labeled spermatozoa were deposited via LAI into the left (CT-Green) and right (CT-Red) uterine horns (n = 4). After ovariohysterectomy (6 hours after insemination), the distributions of green- and red-colored spermatozoa were assessed via tissue section, flushing, and the oviductal contents were also collected. Experiment 3 was designed to test the pregnancy rates in a group of 120 does after LAI using different numbers of spermatozoa (60 and 120 × 10(6) sperm per LAI) and different deposition sites. The results demonstrated that the fluorochromes used in this study did not impair sperm motility or viability. Frozen-thawed goat spermatozoa can migrate transuterinally after LAI, as evidenced by the observations of both CT-Green- and CT-Red-labeled spermatozoa in both uterine horns. Lower numbers of spermatozoa (60 × 10(6)) that are inseminated unilaterally (either ipsilateral or contralateral to the site of ovulation) can efficiently be used for LAI in goats (with a 56.67% pregnancy rate).
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Fructose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(−)-Fructose, tested according to Ph. Eur.