Merck
  • Efficacy of Helicobacter pylori eradication for the 1st line treatment of immune thrombocytopenia patients with moderate thrombocytopenia.
Efficacy of Helicobacter pylori eradication for the 1st line treatment of immune thrombocytopenia patients with moderate thrombocytopenia.
Annals of hematology (2014-12-17)
Hawk Kim, Won-Sik Lee, Kyoo-Hyung Lee, Sung Hwa Bae, Min Kyoung Kim, Young-Don Joo, Dae Young Zang, Jae-Cheol Jo, Sang Min Lee, Je-Hwan Lee, Jung-Hee Lee, Dae-Young Kim, Hun-Mo Ryoo, Myung Soo Hyun, Hyo Jung Kim
ABSTRACT
The practical usefulness of Helicobacter pylori eradication for immune thrombocytopenia (ITP) patients is still controversial. However, some ITP patients respond to H. pylori eradication. We conducted a multi-center, open label, prospective phase II study to define the efficacy and toxicities of H. pylori eradication as the first line treatment for persistent or chronic ITP patients with moderate thrombocytopenia. Patients with persistent or chronic ITP showing moderate thrombocytopenia (30 × 10(9)/L ≤ platelet count ≤ 70 × 10(9)/L) and positive C(13)-urea breath test (UBT) were selected. Medication consisted of lansoprazole 30 mg, amoxicillin 1000 mg, and clarithromycin 500 mg orally twice daily for a week. Complete response (CR) rate at 4 weeks, 3 months, 6 months, 12 months, and maximal response was 19.2, 50.0, 50.0, 26.9, and 65.4%, respectively. Overall response rate (ORR) at 4 weeks, 3 months, 6 months, 12 months, and maximal response was 19.2, 57.7, 65.4, 30.8, and 69.2%, respectively. Median maximal platelet count during the first 3 months was 110 × 10(9)/L (range, 40-274). Median time to CR was 8 weeks (95% CI = 5.429-10.571). Median time to ORR was 4 weeks (95% CI = 1.228-6.772). Only per-protocol population was a response predictor for ORR at 3 months (70.0%, p = 0.054) and maximal ORR (80.0%, p = 0.051), but not for CR at 3 months (60.0%, p = 0.160). Therefore, eradication of H. pylori is an effective and durable first line treatment for persistent or chronic ITP with moderate thrombocytopenia with high ORR and rapid onset in this study.
Product Description
Pimozide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pimozide
Sigma-Aldrich
Astemizole, ≥98% (HPLC)