Merck
  • Inhibition of indoleamine 2,3-dioxygenase activity enhances the anti-tumour effects of a Toll-like receptor 7 agonist in an established cancer model.
Inhibition of indoleamine 2,3-dioxygenase activity enhances the anti-tumour effects of a Toll-like receptor 7 agonist in an established cancer model.
Immunology (2014-10-18)
Hiroyasu Ito, Tatsuya Ando, Yuko Arioka, Kuniaki Saito, Mitsuru Seishima
ABSTRACT
Toll-like receptor (TLR) agonists have been shown to have anti-tumour activity in basic research and clinical studies. However, TLR agonist monotherapy does not sufficiently eliminate tumours. Activation of the innate immune response by TLR agonists is effective at driving adaptive immunity via interleukin-12 (IL-12) or IL-1, but is counteracted by the simultaneous induction of immunosuppressive cytokines and other molecules, including IL-10, transforming growth factor-β, and indoleamine 2,3-dioxygenase (IDO). In the present study, we evaluated the anti-cancer effect of the TLR7 agonist, imiquimod (IMQ), in the absence of IDO activity. The administration of IMQ in IDO knockout (KO) mice inoculated with tumour cells significantly suppressed tumour progression compared with that in wild-type (WT) mice, and improved the survival rate. Moreover, injection with IMQ enhanced the tumour antigen-specific T helper type 1 response in IDO-KO mice with tumours. Combination therapy with IMQ and an IDO inhibitor also significantly inhibited tumour growth. Our results indicated that the enhancement of IDO expression with TLR agonists in cancer treatment might impair host anti-tumour immunity while the inhibition of IDO could enhance the therapeutic efficacy of TLR agonists via the increase of T helper type 1 immune response.
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
1-Methyl-D-tryptophan, 95%
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Tryptophan, European Pharmacopoeia (EP) Reference Standard