Merck
  • A national reconnaissance for selected organic micropollutants in sediments on French territory.
A national reconnaissance for selected organic micropollutants in sediments on French territory.
Environmental science and pollution research international (2014-06-04)
Emmanuelle Vulliet, Alexandra Berlioz-Barbier, Florent Lafay, Robert Baudot, Laure Wiest, Antoine Vauchez, François Lestremau, Fabrizio Botta, Cécile Cren-Olivé
ABSTRACT
To collect a large data set regarding the occurrence of organic substances in sediment, this study presents the examination of 20 micropollutants, as a national survey. The list of target compounds contains two alkylphenols, three polycyclic aromatic hydrocarbons (PAHs) not commonly included in monitoring programmes, six pesticides or metabolites, five pharmaceutical compounds, two hormones, one UV filter and bisphenol A. The selective and sensitive analytical methods, based on quick, easy, cheap, effective, rugged, and safe (QuEChERS) sample preparation followed by liquid chromatography-tandem mass spectrometry (LC-MS/MS) or gas chromatography-time-of-flight mass spectrometry (GC-ToF-MS), allow the quantification at limits comprised between 0.5 and 23 ng/g, depending on the compound. The paper summarizes the analytical results from 154 sampling points. Of the 20 target compounds, 9 were determined at least once, and the sediments contained a maximum of 7 substances. The most frequently detected were PAHs (frequency, 77 %; max., 1,400 ng/g). The pharmaceutical compounds, hormones and pesticides were rarely detected in the samples; the most frequently detected was carbamazepine (frequency, 6 %; max., 31 ng/g). In some cases, the levels of PAHs and bisphenol A exceed the predicted no-effect concentration (PNEC) values.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
3-Ethoxy-1,2-propanediol, 98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%