Merck
  • Galectin-3 induces cell migration via a calcium-sensitive MAPK/ERK1/2 pathway.
Galectin-3 induces cell migration via a calcium-sensitive MAPK/ERK1/2 pathway.
Oncotarget (2014-05-09)
Xiaoge Gao, Vitaly Balan, Guihua Tai, Avraham Raz
ABSTRACT
The presence and level of circulating galectin-3 (Gal-3), a member of the galectin family, is associated with diverse diseases ranging from heart failure, immune disorders to cancer metastasis and serves as a biomarker of diagnosis and treatment response. However, the mechanisms by which exogenous Gal-3 affects pathobiology events remain elusive. In the current study, we found that exogenous Gal-3 slightly delays, while prolonging tyrosine phosphorylation of extracellular signal-regulated kinase 1/2 (ERK1/2) in HeLa cells through a calcium-sensitive and PKC-dependent signaling pathway. The activation was dependent on the sugar-binding properties of Gal-3, since the antagonist lactose could inhibit it. The sugar-binding motif of Gal-3 was required for the activation of ERK1/2. The activation of ERK1/2 was necessary for the initiation and induction of cell migration associated with the phosphorylation of paxillin. All the results presented in this study suggest a novel calcium-sensitive and PKC-dependent pathway through which circulating Gal-3 promotes cell migration and activating the ERK1/2. Taken together, the data depicted here propose a biological function and a target for the diseases' associated circulating Gal-3.
Sigma-Aldrich
1,2-Bis(2-Aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid, 98%
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Sigma-Aldrich
m-3M3FBS, >97% (HPLC)
Sigma-Aldrich
1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid, ≥96.0% (HPLC)